Learn More
Cole-Parmer MEDIA BOTTLE PETG 1000 ML 10/C
Catalog No. NC1964911 Shop All Cole-Parmer Products
Encompass_Preferred
Keep your system free of machining residue or bonding agents. These single-use media bottles with tubing and vent filters are perfect for fluid transfer and storage. Each polypropylene (PP) cap assembly features a platinum-cured silicone gasket for a leakproof system without any machining residue or bonding agents. Hose barb plug on tubing keeps line closed until ready for use. Use the pinch clamp to help control flow into the container or to close off the line. A 0.2 m hydrophobic PTFE filter allows for ventilation while preventing media from escaping in the event of a spill. Rigid dip tube with notch provides easy filling and extraction of media.
Catalog No. NC1964911 Supplier Cole-Parmer Supplier No. 0606143
Keep your system free of machining residue or bonding agents. These single-use media bottles with tubing and vent filters are perfect for fluid transfer and storage. Each polypropylene (PP) cap assembly features a platinum-cured silicone gasket for a leakproof system without any machining residue or bonding agents. Hose barb plug on tubing keeps line closed until ready for use. Use the pinch clamp to help control flow into the container or to close off the line. A 0.2 m hydrophobic PTFE filter allows for ventilation while preventing media from escaping in the event of a spill. Rigid dip tube with notch provides easy filling and extraction of media.