Learn More
Grainger Media Bottle Set 273.05 mm H PK10
Catalog No. 502749420
Encompass_Preferred
The Benchmark Scientific Media Bottle Starter Pack is a set of 6 polypropylene media bottles with leakproof, screw-on caps. The bottles have graduations up to 500mL and are autoclavable up to 121°C. This pack includes (6) 250mL bottles with orange caps, ideal for preparing and storing media, buffers, reagents, and cell cultures. The polypropylene construction is durable, reusable, and resistant to chemicals and heat. With leakproof caps and graduations, these bottles allow accurate and contamination-free preparation and storage. The autoclavable bottles provide a convenient and reusable vessel for sterile media and solutions.
Catalog No. 50-274-9420 Supplier Grainger Supplier No. 14N165
The Benchmark Scientific Media Bottle Starter Pack is a set of 6 polypropylene media bottles with leakproof, screw-on caps. The bottles have graduations up to 500mL and are autoclavable up to 121°C. This pack includes (6) 250mL bottles with orange caps, ideal for preparing and storing media, buffers, reagents, and cell cultures. The polypropylene construction is durable, reusable, and resistant to chemicals and heat. With leakproof caps and graduations, these bottles allow accurate and contamination-free preparation and storage. The autoclavable bottles provide a convenient and reusable vessel for sterile media and solutions.