Learn More
Bellco Glass, Inc. Media Bottle Only,5000mL No Cap or Ring
SDP
Catalog No. 501945798
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. DIMENSIONS: 182mm X 330mm TALL. MADE IN THE USA
Catalog No. 50-194-5798 Supplier Bellco Glass, Inc. Supplier No. 563765000
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. DIMENSIONS: 182mm X 330mm TALL. MADE IN THE USA