Learn More
Bellco Glass, Inc. Media Bttle W/Blue S/Cap,100mL Simax
SDP
Catalog No. 501945812
Encompass_Preferred
Manufacture from Type 1 Class A Borosilicate Glass. MADE IN THE USA Size: 100ml Dimensions: 56x105mm Includes: 45mm polypropylene blue screw cap and pouring ring
Catalog No. 50-194-5812 Supplier Bellco Glass, Inc. Supplier No. 563780100
Manufacture from Type 1 Class A Borosilicate Glass. MADE IN THE USA Size: 100ml Dimensions: 56x105mm Includes: 45mm polypropylene blue screw cap and pouring ring