Learn More
Bellco Glass, Inc. Media Bttle W/Blue S/Cap,10L Simax
SDP
Catalog No. 501945818
Encompass_Preferred
Manufacture from Type 1 Class A Borosilicate Glass. Size: 10l Dimensions: 234x435mm Includes: 45mm polypropylene blue screw cap and pouring ring
Catalog No. 50-194-5818 Supplier Bellco Glass, Inc. Supplier No. 563788100
Manufacture from Type 1 Class A Borosilicate Glass. Size: 10l Dimensions: 234x435mm Includes: 45mm polypropylene blue screw cap and pouring ring