Learn More
Bellco Glass, Inc. Media Bttle W/Blue S/Cap,1L Simax
SDP
Catalog No. 501945815
Encompass_Preferred
Manufacture from Type 1 Class A Borosilicate Glass. Size: 1l Dimensions: 101x203mm Includes: 45mm polypropylene blue screw cap and pouring ring
Catalog No. 50-194-5815 Supplier Bellco Glass, Inc. Supplier No. 563781000
Manufacture from Type 1 Class A Borosilicate Glass. Size: 1l Dimensions: 101x203mm Includes: 45mm polypropylene blue screw cap and pouring ring