Learn More
Bellco Glass, Inc. Media Bttle W/Blue S/Cap,2L Schott
SDP
Catalog No. 501945769
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. . On sizes 100-200mL there is a full capacity line. These bottles include a linerless blue polypropylene cap and pouring ring. DIMENSIONS: 136mm X 260mm TALL
Catalog No. 50-194-5769 Supplier Bellco Glass, Inc. Supplier No. 563702000
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. . On sizes 100-200mL there is a full capacity line. These bottles include a linerless blue polypropylene cap and pouring ring. DIMENSIONS: 136mm X 260mm TALL