Learn More
Bellco Glass, Inc. Media Bttle W/Blue S/Cap,500mL Simax
SDP
Catalog No. 501945814
Encompass_Preferred
Manufacture from Type 1 Class A Borosilicate Glass. Size: 500ml Dimensions: 86x182mm Includes: 45mm polypropylene blue screw cap and pouring ring
Catalog No. 50-194-5814 Supplier Bellco Glass, Inc. Supplier No. 563780500
Manufacture from Type 1 Class A Borosilicate Glass. Size: 500ml Dimensions: 86x182mm Includes: 45mm polypropylene blue screw cap and pouring ring