Learn More
Bellco Glass, Inc. Media Bttle W/Blue S/Cap,50mL Schott
SDP
Catalog No. 501945764
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spots. On sizes 100-200mL there is a full capacity line. These bottles include a linerless blue polypropylene cap and pouring ring. DIMENSIONS: 46mm X 87mm TALL
Catalog No. 50-194-5764 Supplier Bellco Glass, Inc. Supplier No. 563700050
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spots. On sizes 100-200mL there is a full capacity line. These bottles include a linerless blue polypropylene cap and pouring ring. DIMENSIONS: 46mm X 87mm TALL