Learn More
Electron Microscopy Sciences Media Lab Bottles Amber, 125 ml
SDP
Catalog No. 5027090
Encompass_Preferred
Made of Wheaton '500' borosilicate glass to protect light-sensitive samples.
Volume: 125 ml
Catalog No. 50-270-90 Supplier Electron Microscopy Sciences Supplier No. 64210A
Made of Wheaton '500' borosilicate glass to protect light-sensitive samples.
Volume: 125 ml