Learn More
Electron Microscopy Sciences Media Lab Bottles Clear, 125 ml
SDP
Catalog No. 5027091
Encompass_Preferred
Made of Wheaton "400" glass, these graduated bottles have a large opening, no-drip pour lip, and sloping shoulders to facilitate pouring and cleaning. They are widely used for mixing and storage of culture media, fixation and dehydrating materials. Bottles show approximate volume graduations, and have large frosted writing area.
Volume: 125 ml
Catalog No. 50-270-91 Supplier Electron Microscopy Sciences Supplier No. 64210C
Made of Wheaton "400" glass, these graduated bottles have a large opening, no-drip pour lip, and sloping shoulders to facilitate pouring and cleaning. They are widely used for mixing and storage of culture media, fixation and dehydrating materials. Bottles show approximate volume graduations, and have large frosted writing area.
Volume: 125 ml