Learn More
Bellco Glass, Inc. Media Storage Bottle,500mL With 33mm Blk Cap
SDP
Catalog No. 501945745
Encompass_Preferred
Manufactured from Type1 Class B Borosilicate Glass. Large opening and sloping shoulders facilitate pouring and cleaning. Heavier walls prevent breakage of glass. Media storage bottles have white graduations and a marking area. They are supplied with rubber lined 38mm black phenolic screw cap. 12 per case MADE IN THE USA
Catalog No. 50-194-5745 Supplier Bellco Glass, Inc. Supplier No. 563600533
Manufactured from Type1 Class B Borosilicate Glass. Large opening and sloping shoulders facilitate pouring and cleaning. Heavier walls prevent breakage of glass. Media storage bottles have white graduations and a marking area. They are supplied with rubber lined 38mm black phenolic screw cap. 12 per case MADE IN THE USA