Learn More
Alkali Scientific Media Storage Bottles, Polystyrene, VFS, Neck 45mm, 250mL, 1/pk, 24/cs
SDP
Catalog No. 5000662896
Encompass_Preferred
These vacuum bottle top filter systems provide a cost effective and efficient solution for sterilizing and clarifying aqueous biological media. Featuring a 0.22 μm PES (polyethersulfone) membrane, they offer high flow rates, low protein binding, and minimal chemical extractables—ideal for general purpose laboratory filtration where extreme chemical resistance is not required. • High flow rates and throughput • Low protein adsorption and low extractables • 0.22 μm PES membrane for reliable sterilization • Sterile, non pyrogenic, DNase/RNase free • E beam sterilized, SAL = 10⁻⁶ • Individually packaged in sterile bags • Endotoxin free and pyrogen free
Catalog No. 50-006-62896 Supplier Alkali Scientific Supplier No. PSB250
These vacuum bottle top filter systems provide a cost effective and efficient solution for sterilizing and clarifying aqueous biological media. Featuring a 0.22 μm PES (polyethersulfone) membrane, they offer high flow rates, low protein binding, and minimal chemical extractables—ideal for general purpose laboratory filtration where extreme chemical resistance is not required. • High flow rates and throughput • Low protein adsorption and low extractables • 0.22 μm PES membrane for reliable sterilization • Sterile, non pyrogenic, DNase/RNase free • E beam sterilized, SAL = 10⁻⁶ • Individually packaged in sterile bags • Endotoxin free and pyrogen free