Learn More
Description
Medibase C is a clear to slightly hazy, colorless or practically colorless, viscous liquid having a slight characteristic odor. It is slightly hygroscopic. Medibase C is miscible with water and soluble in alcohol. For use as an excipient.
Specifications
Specifications
| Chemical Name or Material | Medibase C (Polyglycol 300 MW) (Liquid) |
| Packaging | Bottle, 32oz.Amber PET Boston Round, 28-410 |
| CAS | 25322-68-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 285 to 315 (average) |
| Quantity | 1 L |
| Percent Purity | 97 to 103 % |
| Grade | Ungraded |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
