Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Medibase C (Polyglycol 300 MW) (Liquid)
SDP

Catalog No. 14186131 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-131 20 L
14-185-955 1 L
14-185-956 4 L
14-185-503 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-186-131 Supplier Medisca Supplier No. 075007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Medibase C is a clear to slightly hazy, colorless or practically colorless, viscous liquid having a slight characteristic odor. It is slightly hygroscopic.

Medibase C is a clear to slightly hazy, colorless or practically colorless, viscous liquid having a slight characteristic odor. It is slightly hygroscopic. Medibase C is miscible with water and soluble in alcohol. For use as an excipient.

Specifications

Chemical Name or Material Medibase C (Polyglycol 300 MW) (Liquid)
Packaging Pail, 6 gallon, White, HDPE, Round
CAS 25322-68-3
Type Drug Standard
Molecular Weight (g/mol) 285 to 315 (average)
Quantity 20 L
Percent Purity 97 to 103 %
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.