Learn More
Selleck Chemical LLC Medicagenic acid S3305-1mg
SDP
Catalog No. 503109784
Encompass_Preferred
Medicagenic acid (Castanogenin) a bioactive triterpenoid pentacyclic glycoside isolated from Herniaria glabra L has low xanthine oxidase collagenase elastase and tyrosinase inhibitory activity
Catalog No. 50-310-9784 Supplier Selleck Chemical LLC Supplier No. S33051MG
Add to Cart
Add to Cart
Medicagenic acid (Castanogenin) a bioactive triterpenoid pentacyclic glycoside isolated from Herniaria glabra L has low xanthine oxidase collagenase elastase and tyrosinase inhibitory activity