Learn More
Medchemexpress LLC Medicagenic acid | 599-07-5 | MFCD02752364 | >98.0% | 488.7 g/mol | C30H48O5 | 10 MG
SDP
Catalog No. 5000289346
Encompass_Preferred
Medicagenic acid is a pentacyclic triterpenoid (castanogenin) isolated from the roots of Herniaria glabra and supplied for laboratory research use. It exhibits fungistatic effects against several plant pathogens and human dermatophytes and shows low inhibitory activity against enzymes including xanthine oxidase, collagenase, elastase, tyrosinase, and cholinesterases.
  • High purity (>98%).
  • Slightly yellow to yellow solid form.
  • Stable when stored as powder at -20°C for up to 3 years and in solvent at -80°C for up to 2 years.
  • Supplied in a 10 mg quantity suitable for small-scale research applications.
  • Includes technical documentation such as a datasheet, certificate of analysis, and MSDS.
Catalog No. 50-002-89346 Supplier Medchemexpress LLC Supplier No. HYN247210MG
Medicagenic acid is a pentacyclic triterpenoid (castanogenin) isolated from the roots of Herniaria glabra and supplied for laboratory research use. It exhibits fungistatic effects against several plant pathogens and human dermatophytes and shows low inhibitory activity against enzymes including xanthine oxidase, collagenase, elastase, tyrosinase, and cholinesterases.
  • High purity (>98%).
  • Slightly yellow to yellow solid form.
  • Stable when stored as powder at -20°C for up to 3 years and in solvent at -80°C for up to 2 years.
  • Supplied in a 10 mg quantity suitable for small-scale research applications.
  • Includes technical documentation such as a datasheet, certificate of analysis, and MSDS.