Learn More
Medchemexpress LLC Medicagenic acid (Castanogenin) | 599-07-5 | MFCD02752364 | 5 MG
SDP
Catalog No. 5000421538
Encompass_Preferred
Medicagenic acid (Castanogenin) is isolated from the roots of *Herniaria glabra*. It exhibits potent fungistatic effects against several plant pathogens and human dermatophytes. It also shows low enzyme inhibitory activities, targeting enzymes such as xanthine oxidase, collagenase, elastase, tyrosinase, and ChE.
  • Isolated from the roots of *Herniaria glabra*.
  • Exhibits potent fungistatic effects against several plant pathogens and human dermatophytes.
  • Shows low enzyme inhibitory activities.
  • Targets enzymes including xanthine oxidase, collagenase, elastase, tyrosinase, and ChE.
Catalog No. 50-004-21538 Supplier Medchemexpress LLC Supplier No. HYN24725MG
Medicagenic acid (Castanogenin) is isolated from the roots of *Herniaria glabra*. It exhibits potent fungistatic effects against several plant pathogens and human dermatophytes. It also shows low enzyme inhibitory activities, targeting enzymes such as xanthine oxidase, collagenase, elastase, tyrosinase, and ChE.
  • Isolated from the roots of *Herniaria glabra*.
  • Exhibits potent fungistatic effects against several plant pathogens and human dermatophytes.
  • Shows low enzyme inhibitory activities.
  • Targets enzymes including xanthine oxidase, collagenase, elastase, tyrosinase, and ChE.