Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Biohazardous Waste Bags, LLDPE Film, Flat Pack
SDP

Catalog No. 14828165 Shop All American Precision Plastics Products
Change view
Click to view available options
Dimensions (L x W):
30 x 36 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
14-828-165 30 x 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-828-165 Supplier Medegen Supplier No. F145
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biohazardous Waste Bags pass ASTMD1709-85, 165 Gram Dart Test.

  • Made from low-density polyethylene
  • Minimize the risk of contamination from biohazardous materials during collection and transport
  • Bags are red with ''Biohazardous Waste/Infectious Waste'' and biohazard symbol imprinted in black ink
  • Individually folded

Specifications

Certifications/Compliance ASTM Dart or Elmendorf, DOT CFR 49 173-197
Color Red, Black
For Use With (Application) Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab
Includes Biohazardous or infectious waste with biohazard symbol
Material LLDPE
Thickness (English) 1.20 mil
Dimensions (L x W) 30 x 36 in.
Length (English) 30 in.
Width (English) 36 in.
Product Type Biohazardous Waste Bag
Capacity (English) 20 to 30 gal.
Capacity (Metric) 75.7 to 113.6 L
Length (Metric) 76.2 cm
Width (Metric) 91.44 cm
Thickness (Metric) 0.030 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.