Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
SDP

Catalog No. 14828164 Shop All American Precision Plastics Products
Change view
Click to view available options
Dimensions (L x W):
30.5 x 41 in.
23 x 23 in.
24 x 30 in.
25 x 34 in.
33 x 40 in.
37 x 50 in.
38 x 45 in.
40 x 45 in.
40 x 46 in.
43 x 48 in.
43 x 55 in.
Capacity (English):
12 to 16 gal.
20 to 30 gal.
30 gal.
33 gal.
40 to 45 gal.
44 gal.
55 gal.
7 to 10 gal.
8 to 12 gal.
Thickness (English):
1.1 mil
1.2 mil
1.3 mil
1.4 mil
1.5 mil
1.75 mil
2 mil
3 mil
Certifications/Compliance:
ASTM Dart
ASTM Dart or Elmendorf, DOT CFR 49 173-197
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Capacity (English) Thickness (English) Certifications/Compliance
14-828-164 40 x 46 in. 40 to 45 gal. 1.2 mil ASTM Dart or Elmendorf, DOT CFR 49 173-197
14-828-335 25 x 34 in. 12 to 16 gal. 1.2 mil ASTM Dart
14-828-326 23 x 23 in. 7 to 10 gal. 1.1 mil ASTM Dart
14-828-85 33 x 40 in. 33 gal. 1.5 mil ASTM Dart
14-828-90 38 x 45 in. 44 gal. 1.75 mil ASTM Dart
14-828-89 38 x 45 in. 44 gal. 1.5 mil ASTM Dart
14-828-327 23 x 23 in. 7 to 10 gal. 1.5 mil ASTM Dart
14-828-96 30.5 x 41 in. 30 gal. 2 mil ASTM Dart
14-828-328 30.5 x 41 in. 30 gal. 1.5 mil ASTM Dart
14-828-329 30.5 x 41 in. 30 gal. 3 mil ASTM Dart
14-828-98 38 x 45 in. 44 gal. 3 mil ASTM Dart
18-023-851 38 x 45 in. 44 gal. 1.2 mil ASTM Dart
14-828-331 43 x 48 in. 55 gal. 2 mil ASTM Dart
14-828-108 43 x 55 in. 55 gal. 1.5 mil ASTM Dart
14-828-109 43 x 55 in. 55 gal. 3 mil ASTM Dart
14-828-158 23 x 23 in. 7 to 10 gal. 1.2 mil ASTM Dart or Elmendorf, DOT CFR 49 173-197
14-828-157 37 x 50 in. 44 gal. 1.5 mil ASTM Dart or Elmendorf, DOT CFR 49 173-197
14-828-156 38 x 45 in. 44 gal. 1.2 mil ASTM Dart or Elmendorf, DOT CFR 49 173-197
14-828-161 24 x 30 in. 8 to 12 gal. 1.2 mil ASTM Dart or Elmendorf, DOT CFR 49 173-197
14-828-159 30.5 x 41 in. 20 to 30 gal. 1.2 mil ASTM Dart or Elmendorf, DOT CFR 49 173-197
14-828-163 37 x 50 in. 44 gal. 1.3 mil ASTM Dart or Elmendorf, DOT CFR 49 173-197
14-827-43B 30.5 x 41 in. 30 gal. 1.2 mil ASTM Dart
14-828-166 40 x 45 in. 40 to 45 gal. 1.2 mil ASTM Dart or Elmendorf, DOT CFR 49 173-197
14-828-336 23 x 23 in. 7 to 10 gal. 1.4 mil ASTM Dart
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Catalog No. 14-828-164 Supplier Medegen Supplier No. F135
Add to Cart
Edge
Add to Cart

These Biohazardous Waste Bags pass ASTMD1709-85, 165 Gram Dart Test.

  • Made from low-density polyethylene
  • Minimize the risk of contamination from biohazardous materials during collection and transport
  • Bags are red with ''Biohazardous Waste/Infectious Waste'' and biohazard symbol imprinted in black ink
  • Individually folded

Specifications

Certifications/Compliance ASTM Dart or Elmendorf, DOT CFR 49 173-197
Color Red, Black
For Use With (Application) Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab
Includes Biohazardous or infectious waste with biohazard symbol
Material LLDPE
Thickness (English) 1.2 mil
Dimensions (L x W) 40 x 46 in.
Length (English) 40 in.
Width (English) 46 in.
Product Type Biohazardous Waste Bag
Capacity (English) 40 to 45 gal.
Capacity (Metric) 151.4 to 170.3 L
Length (Metric) 101.6 cm
Width (Metric) 116.84 cm
Thickness (Metric) 0.0304 mm
Show More Show Less

May NOT be used in CA, FL, PA or for interstate (between state) transport.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.