Learn More
- Home
- Products
- Facility Safety and Maintenance
- Waste Disposal
- Hazardous and Biohazardous Waste Disposal Bags and Containers
- Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll

Catalog No. 1482898
Shop All American Precision Plastics Products


| Catalog No. | Dimensions (L x W) | Capacity (English) | Thickness (English) | Certifications/Compliance |
|---|---|---|---|---|
| 14-828-98 | 38 x 45 in. | 44 gal. | 3 mil | ASTM Dart |
| 14-828-335 | 25 x 34 in. | 12 to 16 gal. | 1.2 mil | ASTM Dart |
| 14-828-326 | 23 x 23 in. | 7 to 10 gal. | 1.1 mil | ASTM Dart |
| 14-828-85 | 33 x 40 in. | 33 gal. | 1.5 mil | ASTM Dart |
| 14-828-90 | 38 x 45 in. | 44 gal. | 1.75 mil | ASTM Dart |
| 14-828-89 | 38 x 45 in. | 44 gal. | 1.5 mil | ASTM Dart |
| 14-828-327 | 23 x 23 in. | 7 to 10 gal. | 1.5 mil | ASTM Dart |
| 14-828-96 | 30.5 x 41 in. | 30 gal. | 2 mil | ASTM Dart |
| 14-828-328 | 30.5 x 41 in. | 30 gal. | 1.5 mil | ASTM Dart |
| 14-828-329 | 30.5 x 41 in. | 30 gal. | 3 mil | ASTM Dart |
| 18-023-851 | 38 x 45 in. | 44 gal. | 1.2 mil | ASTM Dart |
| 14-828-331 | 43 x 48 in. | 55 gal. | 2 mil | ASTM Dart |
| 14-828-108 | 43 x 55 in. | 55 gal. | 1.5 mil | ASTM Dart |
| 14-828-109 | 43 x 55 in. | 55 gal. | 3 mil | ASTM Dart |
| 14-828-158 | 23 x 23 in. | 7 to 10 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-157 | 37 x 50 in. | 44 gal. | 1.5 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-156 | 38 x 45 in. | 44 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-161 | 24 x 30 in. | 8 to 12 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-159 | 30.5 x 41 in. | 20 to 30 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-163 | 37 x 50 in. | 44 gal. | 1.3 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-164 | 40 x 46 in. | 40 to 45 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-827-43B | 30.5 x 41 in. | 30 gal. | 1.2 mil | ASTM Dart |
| 14-828-166 | 40 x 45 in. | 40 to 45 gal. | 1.2 mil | ASTM Dart or Elmendorf, DOT CFR 49 173-197 |
| 14-828-336 | 23 x 23 in. | 7 to 10 gal. | 1.4 mil | ASTM Dart |
Description
- Made from low-density polyethylene
- Minimize the risk of contamination from biohazardous materials during collection and transport
- Bags are red with ''Biohazardous Waste/Infectious Waste'' and biohazard symbol imprinted in black ink
- Individually folded
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | ASTM Dart |
| Color | Red, Black |
| For Use With (Application) | Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab |
| Includes | Biohazardous or infectious waste with biohazard symbol |
| Material | LLDPE |
| Thickness (English) | 3 mil |
| Dimensions (L x W) | 38 x 45 in. |
| Length (English) | 38 in. |
| Width (English) | 45 in. |
| Product Type | Biohazardous Waste Bag |
| Capacity (English) | 44 gal. |
| Capacity (Metric) | 166.6 L |
| Length (Metric) | 96.52 cm |
| Width (Metric) | 114.3 cm |
| Thickness (Metric) | 0.076 mm |
| Show More Show Less |
May NOT be used in CA, FL, PA or for interstate (between state) transport.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.