Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Rolling Cart for Sharps Containers
SDP

Catalog No. 14828325 Shop All American Precision Plastics Products
Change view
Click to view available options
For Use With (Equipment):
For 8 and 16 gal. containers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-828-325 For 8 and 16 gal. containers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-828-325 Supplier Medegen Supplier No. 8790
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rolling Cart for Sharps Containers with locking casters for 8705 and 8716.

  • Features locking casters.
  • For use with Stackable Sharps-Tainers (8705 and 8716).

Specifications

Color White
Includes Locking casters
For Use With (Equipment) For 8 and 16 gal. containers
Material Metal
Product Type Sharps Container Cart
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.