Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Autoclavable Biohazard Waste Bags
GSA_VASDP

Catalog No. 14828334 Shop All American Precision Plastics Products
Change view
Click to view available options
Dimensions (L x W):
6 x 9 in.
9 x 12.5 in.
12 x 15 in.
30 x 36 in.
Color:
Red, Black
Transparent
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color
14-828-334 30 x 36 in. -
22-032197 6 x 9 in. Transparent
14-828-86 9 x 12.5 in. Red, Black
14-828-87 12 x 15 in. Red, Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-828-334 Supplier Medegen Supplier No. X2264
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Autoclavable Biohazard Waste Bags printed with English/Spanish Biohazard and Biohazard Symbol.

  • Print/Label: English/Spanish Biohazard, Biohazard Symbol

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Material Polyethylene
Thickness (English) 3 mil
Dimensions (L x W) 30 x 36 in.
Length (English) 36 in.
Width (English) 30 in.
Product Type Biohazardous Waste Bag
Length (Metric) 90 cm
Width (Metric) 75 cm
Thickness (Metric) 0.08 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.