Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Autoclavable Biohazard Waste Bags
SDP

Catalog No. 1482887 Shop All American Precision Plastics Products
Change view
Click to view available options
Dimensions (L x W):
6 x 9 in.
9 x 12.5 in.
12 x 15 in.
30 x 36 in.
Color:
Red, Black
Transparent
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color
14-828-87 12 x 15 in. Red, Black
22-032197 6 x 9 in. Transparent
14-828-86 9 x 12.5 in. Red, Black
14-828-334 30 x 36 in. -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-828-87 Supplier Medegen Supplier No. 4921
Add to Cart
Edge
Add to Cart

Autoclavable Biohazard Waste Bags printed with English/Spanish Biohazard and Biohazard Symbol.

  • Print/Label: English/Spanish Biohazard, Biohazard Symbol

Specifications

Color Red, Black
Material Polyethylene
Thickness (English) 2 mil
Dimensions (L x W) 12 x 15 in.
Length (English) 15 in.
Width (English) 12 in.
Product Type Biohazardous Waste Bag
Length (Metric) 38 cm
Width (Metric) 30 cm
Thickness (Metric) 0.05 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.