Learn More
PTSOLUTIONS Oxygen Single Stage flow gauge regulator 2-15 LPM
Catalog No. NC9628920
Encompass_Preferred
Chrome-plated brass body. 3000 psi
Catalog No. NC9628920 Supplier PTSOLUTIONS Supplier No. M1540P
Chrome-plated brass body. 3000 psi