Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PixCell Medical™ HemoScreen Analyzer

Catalog No. 23123800
Change view
Click to view available options
Depth (English):
10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English)
23-123-800 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-123-800 Supplier PixCell Medical™ Supplier No. HSFA30219
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PixCell™ Medical HemoScreen offers a true point-of-care solution for testing Complete Blood Count (CBC) with 5-part diff, providing lab-quality precision with a small-sized, simple diagnostic device. HemoScreen is the only CBC with 5-part diff that is FDA-cleared for point-of-care use.

  • Portable, automated hematology instrument used for the quantitative determination of Complete Blood Count (CBC) parameters.
  • Measures 20 standard CBC parameters including 5-part leukocyte differential for venous and capillary whole blood specimens.
  • Makes POC blood testing exceptionally simple using a disposable Sampler and Cartridge pairing (Test Kit).
  • Test Kit includes all necessary reagents and requires no maintenance or calibration.
  • Turnaround time is 5 minutes from test to result with the accuracy and precision of advanced central lab analyzers.
  • For in vitro diagnostic use only.

Specifications

Type Autoread Plus Analyzer w/ Accessories
Depth (English) 10 in.
Depth (Metric) 254 mm
Height (English) 12 in.
Height (Metric) 304.8 mm
Width (English) 7 in.
Width (Metric) 177.8 mm
Voltage 100/240 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.