Learn More
Medicus Health Medical Keyboard Covers, Desktop Computer
SDP
Catalog No. NC2290544
Encompass
Keyboard Covers use protective covers on keyboards to make disinfection easier. Keyboard Covers designed to repel liquid spills, dust, dirt, blood and other hazardous agents. Keyboard Covers are made from high quality Polyurethane, our products provide 24/7 protection. Keyboard Covers fit virtually every make and model. Please provide the make and model number of your keyboard - Found on the underside of your keyboard.Computer Keyboard Skins are custom made per order and non-returnable.
This item is not returnable. View return policy
Catalog No. NC2290544 Supplier Medicus Health Supplier No. 5190M1
Keyboard Covers use protective covers on keyboards to make disinfection easier. Keyboard Covers designed to repel liquid spills, dust, dirt, blood and other hazardous agents. Keyboard Covers are made from high quality Polyurethane, our products provide 24/7 protection. Keyboard Covers fit virtually every make and model. Please provide the make and model number of your keyboard - Found on the underside of your keyboard.Computer Keyboard Skins are custom made per order and non-returnable.