Learn More
Grainger Meret Me Medical Oxygen Cylinder, 670 L, Aluminum, 1 ea
Catalog No. 502782469
Encompass_Preferred
The ME Medical Oxygen Cylinder is a durable, refillable oxygen storage solution engineered for reliable performance. Constructed from high-strength 6061-T6 aluminum alloy, this cylinder is manufactured to meet or exceed DOT 3AL and TC 3ALM requirements, ensuring safety and quality in medical oxygen delivery applications.
  • Constructed from lightweight, high-strength 6061-T6 aluminum alloy
  • Capacity of 670 L with 2,015 PSI filling pressure
  • Features a clear powder coat protective finish for increased longevity
  • Equipped with a green neck to clearly identify contents as oxygen
  • Includes Z-valve and features a 0.750-16 UNF-2B thread size
Catalog No. 50-278-2469 Supplier Grainger Supplier No. 19MN07
The ME Medical Oxygen Cylinder is a durable, refillable oxygen storage solution engineered for reliable performance. Constructed from high-strength 6061-T6 aluminum alloy, this cylinder is manufactured to meet or exceed DOT 3AL and TC 3ALM requirements, ensuring safety and quality in medical oxygen delivery applications.
  • Constructed from lightweight, high-strength 6061-T6 aluminum alloy
  • Capacity of 670 L with 2,015 PSI filling pressure
  • Features a clear powder coat protective finish for increased longevity
  • Equipped with a green neck to clearly identify contents as oxygen
  • Includes Z-valve and features a 0.750-16 UNF-2B thread size