Learn More
Medical Packaging CytoSoft™ Cytology Brush
Catalog No. 22263357
Change view
Click to view available options
Product Type:
Cytology Brush
Cytology Brush Pack
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Sterility
22-263357 Cytology Brush Pack Sterile
22-263358 Cytology Brush Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-263357 Supplier Medical Packaging Supplier No. CYB1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Small, smooth brush fibers create a monolayer of cells that enhance interpretation
  • The CytoSoft nylon brush increases collection of endocervical cells and component material
  • Increased cellular yield reduces the liability associated with inadequate specimens and false negative specimens obscured by blood during sampling
Specifications
Product Type Cytology Brush Pack
Material Nylon
Sterility Sterile
Length (English) 6 in.
Length (Metric) 20.3 cm
For Use With (Application) Used to collect specimens for sexually transmitted disease (STD) testing
Quantity 500 Cs.