Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medical Packaging Pap-Pak™ Kit with the Papette™ Cervical Cell Collector
GSA_VA

Catalog No. 22037749
Change view
Click to view available options
Quantity:
10 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-037749 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-037749 Supplier Medical Packaging Supplier No. CSW1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Wallach Papette collects all cells essential for tracing the early stages of cervical cancer

  • By inserting into the cervical canal and rotating once or twice while applying light pressure, the 8 in. (20cm) sampler collects endocervical, ectocervical, and T-zone cells
  • The soft, flexible polyethylene bristles of the Papette provide enhanced pressure control and prevent internal injury, which can result in smears obscured by blood during sampling

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Full-spectrum Cervical Broom Cell Collector
Sterility Non-sterile
Sample Type Endocervical, ectocervical, and T-zone cells
For Use With (Application) Collects all cells essential for tracing the early stages of cervical cancer
Weight (English) 10 lb.
Weight (Metric) 4.5 kg
Quantity 10 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.