Learn More
Medical Packaging Swab-Pak™ Swabs
GSA_VA
Catalog No. 22281661
Change view
Click to view available options
Tip Type:
Dual End
Single End
Handle Material:
Aluminum
Plastic
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Type Handle Material
22-281661 Single End Aluminum
22-281659 Dual End Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-281661 Supplier Medical Packaging Supplier No. SP7D
Add to Cart
Add to Cart
This item is not returnable. View return policy
Polyester-tipped, general-use swabs
Swabs are contained in a sterile, plastic transport tube with a tamper-evident label
Single swabs have a straight wire shaft on a plastic handle
  • Dual swabs have a plastic shaft
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Handle Material Aluminum
Tip Type Single End
Product Type Swab
Tip Material Dacron
Sterility Sterile
Includes Plastic Tamper-evident labeled transport tube
For Use With (Application) General-use swabs