Learn More
Electron Microscopy Sciences Medical Vial Mailing Boxes, Size: 4-3/4" x 3". Height: 1-3/8"., 1/EA
SDP
Catalog No. 502090617
Encompass_Preferred
The vial mailing boxes are specially designed with reusable wire fasteners and foam padding on the inside for safe and secure transit of standard size medical vials. For for 1cc (ml) through 10cc (ml) medical vials. Size: 4-3/4" x 3". Height: 1-3/8" .
Catalog No. 50-209-0617 Supplier Electron Microscopy Sciences Supplier No. 6154003
The vial mailing boxes are specially designed with reusable wire fasteners and foam padding on the inside for safe and secure transit of standard size medical vials. For for 1cc (ml) through 10cc (ml) medical vials. Size: 4-3/4" x 3". Height: 1-3/8" .