Learn More
Medicus Health Medication Drop Box, Key Lock
SDP
Catalog No. NC3372620
Encompass
Medication Security Drop Box safely stores unused, expired or discontinued drugs being returned to the pharmacy
∙ Medications are deposited via the 12" W x 9"D front top opening on the front of the unit and retrieved via the bottom keyed compartment
∙ Designed to accept large volumes of vials, syringes, ampules, unit dose packaging and other medications
∙ Box can also be used to safely store keys, blood samples or lab work that you need to stage in a safe, locked receptacle until the proper personnel can retrieve it
∙ Pick-resistant locks that are coded in a non-repeating sequence for increased security to help prevent medication theft
∙ Top deposit slot has a metal anti-theft baffle
∙ Made from powder coated white, 18-gauge steel which provides durable construction for demanding hospital environments
∙ Dimensions: 17"W x 14.5"D x 20"H
Catalog No. NC3372620 Supplier Medicus Health Supplier No. 2493M1
Medication Security Drop Box safely stores unused, expired or discontinued drugs being returned to the pharmacy
∙ Medications are deposited via the 12" W x 9"D front top opening on the front of the unit and retrieved via the bottom keyed compartment
∙ Designed to accept large volumes of vials, syringes, ampules, unit dose packaging and other medications
∙ Box can also be used to safely store keys, blood samples or lab work that you need to stage in a safe, locked receptacle until the proper personnel can retrieve it
∙ Pick-resistant locks that are coded in a non-repeating sequence for increased security to help prevent medication theft
∙ Top deposit slot has a metal anti-theft baffle
∙ Made from powder coated white, 18-gauge steel which provides durable construction for demanding hospital environments
∙ Dimensions: 17"W x 14.5"D x 20"H