Learn More
McKesson General Medical Medication Suspending Vehicle Ora-Plus 16 oz. Unflavored
Catalog No. NC1775867
Encompass
Unique suspending vehicle used to simplify the process in the extemporaneous compounding of oral suspensions. Medicated powder can be incorporated into Ora-Plus to form elegant, uniform and physically stable suspensions.
This item is not returnable. View return policy
Catalog No. NC1775867 Supplier McKesson General Medical Supplier No. 636819
Unique suspending vehicle used to simplify the process in the extemporaneous compounding of oral suspensions. Medicated powder can be incorporated into Ora-Plus to form elegant, uniform and physically stable suspensions.