Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ SafeBasics™ Lab Gowns

Catalog No. 19910554 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-910-554 Small
19-910-555 Medium
19-910-556 Large
19-910-557 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-910-554 Supplier Medicom™ Supplier No. 8016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Added protection from cross-contamination.

  • Increased breathability with 3-ply SMS fabric
  • Comfortable fit with knitted cuffs and collar
  • Front snap closure
  • Fluid resistant material
  • 10 per bag, 5 bags per case

Specifications

Size Small
Product Type Single-use Lab Coat
For Use With (Application) For protection from cross contamination
Gender Unisex
No. of Pockets 2
Material SMS
Color Blue
Closure Type Snaps
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Latex Free Yes
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.