Learn More
MEDILUMINE INC MediLumine Inc. 1 Rack for PD 10 Desalting Columns
Catalog No. NC3968182
Encompass_Preferred
Reusable and washable PD-10 Column Rack is designed to fit up to 8 PD-10 size exclusion columns for sample purification. This column rack is a replacement for the disposable cardboard PD-10 column rack, which cannot be washed or decontaminated.
Catalog No. NC3968182 Supplier MEDILUMINE INC Supplier No. MDLPD10
Reusable and washable PD-10 Column Rack is designed to fit up to 8 PD-10 size exclusion columns for sample purification. This column rack is a replacement for the disposable cardboard PD-10 column rack, which cannot be washed or decontaminated.