Learn More
MEDILUMINE INC MediLumine Inc. Bradykinin B1R Agonist, 1 mg
Catalog No. NC3968110
Encompass_Preferred
Metabolically stable agonist of Bradykinin B1 receptor for in vitro & in vivo preclinical research.Temporary permeabilization of the blood-tumour barrier in brain cancer, biomarker research in fields of diabetes and alzheimer’s research.
Catalog No. NC3968110 Supplier MEDILUMINE INC Supplier No. B1111
Metabolically stable agonist of Bradykinin B1 receptor for in vitro & in vivo preclinical research.Temporary permeabilization of the blood-tumour barrier in brain cancer, biomarker research in fields of diabetes and alzheimer’s research.