Learn More
MEDILUMINE INC MediLumine Inc. D-Luciferin Potassium Salt, 1 g
Catalog No. NC3968102
Encompass_Preferred
Highly pure D-Luciferin potassium salt. This potassium salt is a water soluble form of D-Luciferin. D-Luciferin is a common bioluminescent substrate used for imaging of firefly luciferase expression with peak chemiluminescent emission at 560 nm. The ATP dependant luciferin oxidation is catalyzed by the Luciferase enzyme encoded by luc gene.
Catalog No. NC3968102 Supplier MEDILUMINE INC Supplier No. 222PS
Highly pure D-Luciferin potassium salt. This potassium salt is a water soluble form of D-Luciferin. D-Luciferin is a common bioluminescent substrate used for imaging of firefly luciferase expression with peak chemiluminescent emission at 560 nm. The ATP dependant luciferin oxidation is catalyzed by the Luciferase enzyme encoded by luc gene.