Learn More
MEDILUMINE INC MediLumine Inc. Fenestra VC, 2.5 ml
Catalog No. NC3968126
Encompass_Preferred
Fenestra VC contrast agent is an iodinated nanoemulsion used for micro-CT imaging of soft tissues. It is a specialized version of Fenestra LC in which the surface of the lipid emulsion particles is modified with DSPE-PEG which blocks the interaction with APO-E thus enabling preclinical angiography by delaying the targeted uptake by hepatocytes of the lipid spheres. Like its liver-selective counterpart, Fenestra VC is eventually metabolized and eliminated through the hepatobiliary system enabling multiple injection longitudinal studies.
Catalog No. NC3968126 Supplier MEDILUMINE INC Supplier No. VC131
Fenestra VC contrast agent is an iodinated nanoemulsion used for micro-CT imaging of soft tissues. It is a specialized version of Fenestra LC in which the surface of the lipid emulsion particles is modified with DSPE-PEG which blocks the interaction with APO-E thus enabling preclinical angiography by delaying the targeted uptake by hepatocytes of the lipid spheres. Like its liver-selective counterpart, Fenestra VC is eventually metabolized and eliminated through the hepatobiliary system enabling multiple injection longitudinal studies.