Learn More
MEDILUMINE INC MediLumine Inc. In Vivo Gripper
Catalog No. NC3968131
Encompass_Preferred
The In Vivo Grip Platform is an engineered solution designed specifically to address the nuances of rodent research by improving handling precision and subject welfare. Featuring a patented perforated design, the platform allows mice to engage their natural gripping instinct, which significantly reduces animal stress and anxiety compared to traditional handling techniques. This reduction in stress not only promotes superior animal welfare but also enhances data integrity by ensuring more reliable physiological and behavioral responses. Furthermore, the platform increases handler safety by providing a secure interface that minimizes the risk of bites and scratches. By integrating this tool into your laboratory workflow, you can improve your handling proficiency and focus on high-quality research outcomes through a more humane, efficient, and standardized methodology.
Catalog No. NC3968131 Supplier MEDILUMINE INC Supplier No. MDLIVG001
The In Vivo Grip Platform is an engineered solution designed specifically to address the nuances of rodent research by improving handling precision and subject welfare. Featuring a patented perforated design, the platform allows mice to engage their natural gripping instinct, which significantly reduces animal stress and anxiety compared to traditional handling techniques. This reduction in stress not only promotes superior animal welfare but also enhances data integrity by ensuring more reliable physiological and behavioral responses. Furthermore, the platform increases handler safety by providing a secure interface that minimizes the risk of bites and scratches. By integrating this tool into your laboratory workflow, you can improve your handling proficiency and focus on high-quality research outcomes through a more humane, efficient, and standardized methodology.