Learn More
MEDILUMINE INC MediLumine Inc. Mbio Cycluc1 Injectable Luciferin Analog, 5 vials with 5mg/ml, 0.45ml each
Catalog No. NC3968143
Encompass_Preferred
Mbio Cycluc1 is ready to use luciferin analog ideal for in vivo imaging. It can be administered via intravenous (i.v.), subcutaneous (s.c.), or intraperitoneal (i.p.) injections. The product gives a luminescence peak at 620 nm (orange to red window) when used with the standard firefly luciferase reporter system. This enables deeper and more sensitive tissue imaging due to the reduced absorption and scattering of light compared to photonic emissions of D-Luciferin. The analog also provides stronger emissions for in vitro assays compared to the use of D-Luciferin.
Catalog No. NC3968143 Supplier MEDILUMINE INC Supplier No. BIO545
Mbio Cycluc1 is ready to use luciferin analog ideal for in vivo imaging. It can be administered via intravenous (i.v.), subcutaneous (s.c.), or intraperitoneal (i.p.) injections. The product gives a luminescence peak at 620 nm (orange to red window) when used with the standard firefly luciferase reporter system. This enables deeper and more sensitive tissue imaging due to the reduced absorption and scattering of light compared to photonic emissions of D-Luciferin. The analog also provides stronger emissions for in vitro assays compared to the use of D-Luciferin.