Learn More
MEDILUMINE INC MediLumine Inc. Mouse Malocclusion Holder
Catalog No. NC3968133
Encompass_Preferred
Allows user to control the trimming of the teeth of a mouse without injury. Suction cups allow the holder to be attached to the inside of the glass sash of a hood.Once the teeth are behind the cotton swab you can easily trim the teeth with a trimmer
Catalog No. NC3968133 Supplier MEDILUMINE INC Supplier No. MH111
Allows user to control the trimming of the teeth of a mouse without injury. Suction cups allow the holder to be attached to the inside of the glass sash of a hood.Once the teeth are behind the cotton swab you can easily trim the teeth with a trimmer