Learn More
MEDILUMINE INC MediLumine Inc. Vascupaint™ clear kit with one bottle of clear silicone (200 ml), one bottle of diluent (200 ml), and one bottle of catalyst (20 ml)
Catalog No. NC3968118
Encompass_Preferred
Vascupaint™ Clear is a lead-free, silicone-based injection compound specifically engineered for researchers requiring bespoke imaging solutions. This formulation allows for the seamless integration of user-defined, oil-soluble fluorescent dyes, providing total control over excitation and emission profiles for complex vascular mapping. Designed with a lower viscosity than traditional lead-based media, it ensures effortless perfusion of microvasculature in both non-surviving organisms and human organs for ex vivo 3D analysis. Following alcohol-based tissue clearing, Vascupaint™ Clear provides a highly transparent matrix, ideal for high-resolution 3D imaging using fluorescence or brightfield microscopy.
Catalog No. NC3968118 Supplier MEDILUMINE INC Supplier No. MDL123
Vascupaint™ Clear is a lead-free, silicone-based injection compound specifically engineered for researchers requiring bespoke imaging solutions. This formulation allows for the seamless integration of user-defined, oil-soluble fluorescent dyes, providing total control over excitation and emission profiles for complex vascular mapping. Designed with a lower viscosity than traditional lead-based media, it ensures effortless perfusion of microvasculature in both non-surviving organisms and human organs for ex vivo 3D analysis. Following alcohol-based tissue clearing, Vascupaint™ Clear provides a highly transparent matrix, ideal for high-resolution 3D imaging using fluorescence or brightfield microscopy.