Learn More
MEDILUMINE INC MediLumine Inc. Vascupaint™ green kit with one bottle of green silicone (200 ml), one bottle of diluent (200 ml), and one bottle of catalyst (20 ml)
Catalog No. NC3968122
Encompass_Preferred
Vascupaint™ Green is a lead-free, silicone-based injection compound specifically optimized for tissue clearing protocols and high-resolution fluorescence imaging. Its low-viscosity formulation ensures deep perfusion into delicate microvasculature structures in both human organs and animal models, outperforming traditional lead-based alternatives in ease of use. Once injected, the compound provides robust opacification for visual inspection and 3D mapping.Engineered for stability during alcohol-based clearing, Vascupaint™ Green retains its intense fluorescent signal, making it the ideal choice for researchers utilizing brightfield or fluorescence microscopy to generate detailed 3D vascular casts.
Catalog No. NC3968122 Supplier MEDILUMINE INC Supplier No. MDL122
Vascupaint™ Green is a lead-free, silicone-based injection compound specifically optimized for tissue clearing protocols and high-resolution fluorescence imaging. Its low-viscosity formulation ensures deep perfusion into delicate microvasculature structures in both human organs and animal models, outperforming traditional lead-based alternatives in ease of use. Once injected, the compound provides robust opacification for visual inspection and 3D mapping.Engineered for stability during alcohol-based clearing, Vascupaint™ Green retains its intense fluorescent signal, making it the ideal choice for researchers utilizing brightfield or fluorescence microscopy to generate detailed 3D vascular casts.