Learn More
MEDILUMINE INC MediLumine Inc. Vascupaint™ yellow kit with one bottle of yellow silicone (200 ml), one bottle of diluent (200 ml), and one bottle of catalyst (20 ml)
Catalog No. NC3968121
Encompass_Preferred
Vascupaint™ Yellow is a high-performance, lead-free silicone injection compound specifically optimized for micro-CT imaging of microvascular structures. This yellow-pigmented formula provides exceptional X-ray attenuation, allowing for high-contrast 3D visualization of vascular networks following alcohol-based tissue clearing or within opaque samples. As a low-viscosity alternative to traditional lead-based media, Vascupaint™ Yellow ensures effortless and complete perfusion of non-surviving organisms and human organs for precise ex vivo vascular cast analysis. While the Vascupaint™ line includes green, blue, and clear options for fluorescence or custom preparations, the Yellow formulation is the primary choice for researchers requiring maximum fidelity and structural detail in computed tomography.
Catalog No. NC3968121 Supplier MEDILUMINE INC Supplier No. MDL121
Vascupaint™ Yellow is a high-performance, lead-free silicone injection compound specifically optimized for micro-CT imaging of microvascular structures. This yellow-pigmented formula provides exceptional X-ray attenuation, allowing for high-contrast 3D visualization of vascular networks following alcohol-based tissue clearing or within opaque samples. As a low-viscosity alternative to traditional lead-based media, Vascupaint™ Yellow ensures effortless and complete perfusion of non-surviving organisms and human organs for precise ex vivo vascular cast analysis. While the Vascupaint™ line includes green, blue, and clear options for fluorescence or custom preparations, the Yellow formulation is the primary choice for researchers requiring maximum fidelity and structural detail in computed tomography.