Learn More
Cayman Chemical MedIMchaIn Saturated Faty 50mg
SDP
Catalog No. 502939796
Encompass_Preferred
A mixture of five medium-chain saturated fatty acid methyl esters; intended for use as a reference standard for the detection of medium-chain saturated fatty acids in various sample types by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation
Catalog No. 50-293-9796 Supplier Cayman Chemical Supplier No. 2865050 MG
A mixture of five medium-chain saturated fatty acid methyl esters; intended for use as a reference standard for the detection of medium-chain saturated fatty acids in various sample types by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation