Learn More
Active Motif MeDIP
SDP
Catalog No. NC1456479
Encompass
The MeDIP Kit is designed to immunoprecipitate and enrich for single-stranded DNA fragments containing 5-methylcytosine (5-mC). The MeDIP Kit contains a monoclonal 5-methylcytidine antibody, a bridging antibody and the necessary buffers to perform methylated DNA immunoprecipitation (MeDIP). The affinity of the antibody enables separation of 5-mC DNA from 5-hmC DNA using single-stranded input DNA. Active Motif's fast, magnetic protocol has been streamlined to minimize the number of wash and incubation steps, saving you valuable time. The kit also includes MseI digested human genomic DNA and PCR primers that can be used to verify the efficiency of the enrichment. Additionally, the kit contains a bar magnet for easy separation and elution of the enriched methylated DNA.
Catalog No. NC1456479 Supplier Active Motif Supplier No. 55009
The MeDIP Kit is designed to immunoprecipitate and enrich for single-stranded DNA fragments containing 5-methylcytosine (5-mC). The MeDIP Kit contains a monoclonal 5-methylcytidine antibody, a bridging antibody and the necessary buffers to perform methylated DNA immunoprecipitation (MeDIP). The affinity of the antibody enables separation of 5-mC DNA from 5-hmC DNA using single-stranded input DNA. Active Motif's fast, magnetic protocol has been streamlined to minimize the number of wash and incubation steps, saving you valuable time. The kit also includes MseI digested human genomic DNA and PCR primers that can be used to verify the efficiency of the enrichment. Additionally, the kit contains a bar magnet for easy separation and elution of the enriched methylated DNA.