Learn More
MSC Medique Digital Thermometer, Oral, Case, ASTM 1112, 1 ea
Catalog No. 5000673762
Encompass
This oral digital thermometer provides quick, easy-to-read temperatures in 60 seconds. It meets ASTM 1112 standard for accuracy and features a temperature range from 90°F to 109°F.
  • Large, easy-to-read LCD display
  • Memory recalls last reading
  • Water resistant
  • Auto power off to conserve battery
  • Includes 1.55V battery
  • Integral carrying case
  • One button operation
  • Includes 5 probe covers
Catalog No. 50-006-73762 Supplier MSC Supplier No. 45908530
This oral digital thermometer provides quick, easy-to-read temperatures in 60 seconds. It meets ASTM 1112 standard for accuracy and features a temperature range from 90°F to 109°F.
  • Large, easy-to-read LCD display
  • Memory recalls last reading
  • Water resistant
  • Auto power off to conserve battery
  • Includes 1.55V battery
  • Integral carrying case
  • One button operation
  • Includes 5 probe covers