Learn More
MSC MEDIQUE FIRST-AID REFILL
Catalog No. 5000792520
Encompass
Medique Antiseptic Wipes are individual towelettes saturated with 0.13% w/v Benzalkonium Chloride Solution. They serve as a first aid antiseptic, helping to prevent infection in minor cuts, scrapes, and burns, and can also be used for cleaning the face, hands, and body.
  • Saturated with 0.13% w/v Benzalkonium Chloride
  • Each box contains 100 wipes
  • First aid antiseptic for minor cuts, scrapes, and burns
  • Can be used for cleaning face, hands, and body
Catalog No. 50-007-92520 Supplier MSC Supplier No. 45909603
Medique Antiseptic Wipes are individual towelettes saturated with 0.13% w/v Benzalkonium Chloride Solution. They serve as a first aid antiseptic, helping to prevent infection in minor cuts, scrapes, and burns, and can also be used for cleaning the face, hands, and body.
  • Saturated with 0.13% w/v Benzalkonium Chloride
  • Each box contains 100 wipes
  • First aid antiseptic for minor cuts, scrapes, and burns
  • Can be used for cleaning face, hands, and body