Learn More
Medisca™ Medisca MAZ™ Standard Mixing Container
GREENER_CHOICESDP
Catalog No. 17985477 Shop All Medisca Products
Change view
Click to view available options
Size:
300 mL
400 mL
1 L
2 L
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-985-477 300 mL
17-985-476 400 mL
17-985-475 1 L
17-985-473 2 L
17-985-474 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-985-477 Supplier Medisca™ Supplier No. 676502
Add to Cart
Add to Cart
Standard mixing container for all mixing and deaeration procedures in MAZ mixer units.
  • Designed for all mixing and deaeration operations
  • Reusable container for repeated use
Features:
  • Ensures efficient mixing and deaerating performance
  • Durable for regular laboratory use
Note:
  • Not dishwasher safe
  • May need replacement after extended use
Specifications
Product Type Mixing Container
For Use With (Equipment) KK-300SS
Size 300 mL